SY3H3D676BDE Synthonix Corporation

2-Amino-N-(5-chloropyridin-2-yl)-5-methoxybenzamide

  • MDL number MFCD12841614
  • Cas Number : 280773-17-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/56756.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/56756.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart