SY3H3D676BDF Synthonix Corporation

2-(4-Aminophenyl)-1H-benzimidazol-5-amine

  • MDL number No MDL found
  • Cas Number : 7621-86-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/56758.html

Safety Information

signalword Warning
pcodes P201, P202, P264, P280, P405
MSDS https://synthonix.com/56758.html
pictograms GHS06, GHS07
hstatements H315, H319, H341
Availability details for
Quantity
  • Availability
  • Add to Cart