SY3H3D676BE0 Synthonix Corporation

2-(3-Aminophenyl)-1H-benzo[d]imidazol-6-amine

  • MDL number MFCD13874743
  • Cas Number : 13676-49-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form White to light-yellow powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/56759.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/56759.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart