SY3H3D676BE3 Synthonix Corporation

4-(4-Aminophenoxy)-N-methylpicolinamide

  • MDL number MFCD08692047
  • Cas Number : 284462-37-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Crystal - Powder
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/56806.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/56806.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart