SY3H3D676BE6 Synthonix Corporation

4-Acetoxy-3-(cyclopropylmethoxy)benzoic acid

  • MDL number MFCD29075655
  • Cas Number : 1883347-32-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/56887.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/56887.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart