SY3H3D676BE7 Synthonix Corporation

2-((6-Amino-1-methyl-2-oxo-1,2-dihydroquinolin-3-yl)oxy)-N-methylacetamide

  • MDL number MFCD31698162
  • Cas Number : 2216750-84-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/56902.html

Safety Information

signalword Warning
pcodes P280
MSDS https://synthonix.com/56902.html
pictograms GHS07
hstatements H302, H312, H332
Availability details for
Quantity
  • Availability
  • Add to Cart