SY3H3D676BF2 Synthonix Corporation

(2-Amino-5-bromophenyl)(2-fluorophenyl)methanone

  • MDL number MFCD00038380
  • Cas Number : 1479-58-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form White to off-white to yellow powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/57376.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/57376.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart