SY3H3D676BF8 Synthonix Corporation

2-(3-Amino-4-chlorobenzoyl)benzoic acid

  • MDL number MFCD00007672
  • Cas Number : 118-04-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Light-yellow to yellow powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/57485.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/57485.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart