SY3H3D676BFD Synthonix Corporation

(2-Amino-9H-fluoren-9-yl)methanol

  • MDL number MFCD24646252
  • Cas Number : 162021-14-9
  • Molecular Weight : 211.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Pale-yellow to Yellow-brown Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/57606.html

Safety Information

signalword Warning
pcodes P261,P305,P351,P338
MSDS https://synthonix.com/57606.html
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart