SY3H3D676C00 Synthonix Corporation

2-Amino-N-(o-tolyl)benzamide

  • MDL number MFCD00578802
  • Cas Number : 4943-85-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/57636.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/57636.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart