SY3H3D676C03 Synthonix Corporation

(S)-5-(Aminomethyl)-3-(3-fluoro-4-morpholinophenyl)oxazolidin-2-one

  • MDL number MFCD18379308
  • Cas Number : 168828-90-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/57783.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/57783.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart