SY3H3D676C05 Synthonix Corporation

(2R,3R,4R,5R)-2-(Acetoxymethyl)-5-(4-amino-2-oxo-1,3,5-triazin-1(2H)-yl)tetrahydrofuran-3,4-diyl diacetate

  • MDL number MFCD11113159
  • Cas Number : 10302-78-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/57786.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/57786.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart