SY3H3D676C08 Synthonix Corporation

(2R,3R,4S,5S,6S)-2-(Acetoxymethyl)-6-hydroxytetrahydro-2H-pyran-3,4,5-triyl triacetate

  • MDL number MFCD08274522
  • Cas Number : 22860-22-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form White to light-yellow solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/57906.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/57906.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart