SY3H3D676C1C Synthonix Corporation

(S)-2-Amino-1,1-diphenylpropan-1-ol

  • MDL number No MDL found
  • Cas Number : 78603-91-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/58294.html

Safety Information

MSDS https://synthonix.com/58294.html
Availability details for
Quantity
  • Availability
  • Add to Cart