SY3H3D676C24 Synthonix Corporation

1-[2-Amino-1-(4-methoxyphenyl)ethyl]cyclohexanol Hydrochloride

  • MDL number MFCD06658143
  • Cas Number : 130198-05-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Crystal - Powder
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/58539.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/58539.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart