SY3H3D676C27 Synthonix Corporation

N-(5-Amino-2-methylphenyl)-4-(3-pyridyl)-2-pyrimidineamine

  • MDL number MFCD09028125
  • Cas Number : 152460-10-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form Crystal - Powder / Colour: Slightly pale yellow - Yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/58650.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/58650.html
pictograms GHS07
hstatements H302, H315, H319, H332, H335
Availability details for
Quantity
  • Availability
  • Add to Cart