SY3H3D676C2B Synthonix Corporation

(R)-2-((4-Aminophenethyl)amino)-1-phenylethanol

  • MDL number MFCD28125625
  • Cas Number : 391901-45-4
  • Molecular Weight : 256.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Light-yellow to yellow powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/58737.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/58737.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart