SY3H3D676C3B Synthonix Corporation

(3-(Adamantan-1-yl)-4-methoxyphenyl)boronic acid

  • MDL number MFCD08056355
  • Cas Number : 459423-32-6
  • Molecular Weight : 286.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Referigrator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/59055.html

Safety Information

signalword Warning
pcodes P261,P305,P351,P338
MSDS https://synthonix.com/59055.html
pictograms GHS07
hstatements H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart