SY3H3D676C49 Synthonix Corporation

2-(4-aminophenyl)-2-methylpropanenitrile

  • MDL number MFCD09909912
  • Cas Number : 115279-57-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerator
Physical Form Pale-yellow to Yellow-brown Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/9670.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/9670.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart