SY3H3D676C53 Synthonix Corporation

4-Bromo-3,5-dimethylpyrazole

  • MDL number MFCD00005242
  • Cas Number : 3398-16-1
  • Molecular Weight : 175.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/39180.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/39180.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart