SY3H3D676C5B Synthonix Corporation

2-Bromo-1-chloro-4-fluorobenzene

  • MDL number MFCD00061120
  • Cas Number : 201849-15-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form Clear liquid - Solid / Colour: Colorless - Yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/39223.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/39223.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart