SY3H3D676C83 Synthonix Corporation

1-Bromo-3-chloro-2-methylbenzene

  • MDL number MFCD00061121
  • Cas Number : 62356-27-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form Clear liquid / Colour: Colorless - Pale yellow red
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/39372.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/39372.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart