SY3H3D676C96 Synthonix Corporation

2-Bromo-2-(2-chlorophenyl)acetic acid

  • MDL number MFCD03428513
  • Cas Number : 29270-30-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Crystal - Powder
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/39471.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/39471.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart