SY3H3D676CB0 Synthonix Corporation

3-(Benzo[d][1,3]dioxol-5-yl)acrylic acid

  • MDL number MFCD00005837
  • Cas Number : 2373-80-0
  • Molecular Weight : 192.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Crystal - Powder / Colour: White - Slightly pale yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/39641.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/39641.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart