SY3H3D676CBD Synthonix Corporation

N-((tert-Butoxycarbonyl)oxy)benzimidoyl cyanide

  • MDL number MFCD00064192
  • Cas Number : 58632-95-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form Crystal - Powder / Colour: White - Pale-yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/39702.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/39702.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart