SY3H3D676CC0 Synthonix Corporation

1,2-Bis(2-nitrophenyl)ethane

  • MDL number MFCD00024296
  • Cas Number : 16968-19-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Crystal - Powder / Colour: White - Yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/39721.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/39721.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart