SY3H3D676CC4 Synthonix Corporation

4-Bromo-2,2-diphenylbutyronitrile

  • MDL number MFCD00001845
  • Cas Number : 39186-58-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Crystal - Powder
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/39736.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/39736.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart