SY3H3D676CC5 Synthonix Corporation

4-Butyl-1,1'-Biphenyl

  • MDL number MFCD00100665
  • Cas Number : 37909-95-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form Clear liquid / Colour: Colorless - Slightly pale yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/39739.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/39739.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart