SY3H3D676CC6 Synthonix Corporation

4-(trans-4-Butylcyclohexyl)benzoic acid

  • MDL number MFCD06658178
  • Cas Number : 83626-35-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Crystal - Powder / Colour: White - Almost white
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/39742.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/39742.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart