SY3H3D676CC8 Synthonix Corporation

(2R,3R)-2,3-Bis((4-methoxybenzoyl)oxy)succinic acid

  • MDL number MFCD02682986
  • Cas Number : 50583-51-2
  • Molecular Weight : 418.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Crystal - Powder / Colour: White - Very pale yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/39752.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/39752.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart