SY3H3D676CC9 Synthonix Corporation

(2S,3S)-2,3-Bis((4-methoxybenzoyl)oxy)succinic acid

  • MDL number MFCD07368366
  • Cas Number : 191605-10-4
  • Molecular Weight : 418.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form White Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/39753.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/39753.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart