SY3H3D676CF8 Synthonix Corporation

4-Bromo-3,3,4,4-tetrafluorobut-1-ene

  • MDL number MFCD00042189
  • Cas Number : 18599-22-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Colorless to light-yellow liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/40017.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/40017.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart