SY3H3D676D01 Synthonix Corporation

2-BROMO-THIAZOLE-5-CARBOXAMIDE

  • MDL number MFCD09998942
  • Cas Number : 848499-31-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/40041.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/40041.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart