SY3H3D676D0A Synthonix Corporation

(4-Bromothiophen-2-yl)boronic acid

  • MDL number No MDL found
  • Cas Number : 499769-92-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/40109.html

Safety Information

MSDS https://synthonix.com/40109.html
Availability details for
Quantity
  • Availability
  • Add to Cart