SY3H3D676D16 Synthonix Corporation

4-Bromo-1,5-dimethyl-1H-1,2,3-triazole

  • MDL number MFCD06242922
  • Cas Number : 885877-41-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/40194.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/40194.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart