SY3H3D676D1A Synthonix Corporation

2-Bromo-3-methoxypropanoic acid

  • MDL number MFCD13185991
  • Cas Number : 65090-78-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/40233.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/40233.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart