SY3H3D676D2B Synthonix Corporation

1-Bromo-4-methoxybutane

  • MDL number MFCD00037102
  • Cas Number : 4457-67-4
  • Molecular Weight : 167.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Clear liquid / Colour: Colorless - Pale-yellow
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/40422.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/40422.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart