SY3H3D676D35 Synthonix Corporation

2-Bromo-3-chloro-5-nitropyridine

  • MDL number MFCD08061302
  • Cas Number : 22353-41-9
  • Molecular Weight : 237.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/40459.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/40459.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart