SY3H3D676D44 Synthonix Corporation

3-Bromo-2,5-dichloropyridine

  • MDL number MFCD06659484
  • Cas Number : 138006-41-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form Crystal - Powder / Colour: White - Yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/40505.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/40505.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart