SY3H3D676D8F Synthonix Corporation

3-Bromo-5-nitropyridin-2-amine

  • MDL number MFCD00955628
  • Cas Number : 15862-31-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Pale-yellow to Yellow-brown Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/40795.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/40795.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart