SY3H3D676DA7 Synthonix Corporation

2-Bromo-5-(methylthio)pyrazine

  • MDL number MFCD09753145
  • Cas Number : 1049026-49-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form Pale-yellow to Yellow-brown Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/40873.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/40873.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart