SY3H3D676DAA Synthonix Corporation

5-Bromo-6-methylpyrimidine-2,4-diol

  • MDL number MFCD12964069
  • Cas Number : 15018-56-1
  • Molecular Weight : 205.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/40891.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/40891.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart