SY3H3D676DAD Synthonix Corporation

5-Bromo-6-hydroxy-2-methylpyrimidin-4(1H)-one

  • MDL number MFCD18427081
  • Cas Number : 4722-76-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/40897.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/40897.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart