SY3H3D676DBA Synthonix Corporation

5-Bromo-6-methylpyrimidin-2(1H)-one

  • MDL number MFCD21608032
  • Cas Number : 69849-34-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Referigrator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/40923.html

Safety Information

signalword Warning
pcodes P261, P280
MSDS https://synthonix.com/40923.html
pictograms GHS07
hstatements H302, H315, H320, H335
Availability details for
Quantity
  • Availability
  • Add to Cart