SY3H3D676DBE Synthonix Corporation

4-Bromo-5-(bromomethyl)-2-methylthiazole

  • MDL number MFCD28902472
  • Cas Number : 1799412-37-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form White to off-white to red solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/40988.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/40988.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart