SY3H3D676DE9 Synthonix Corporation

3-Bromo-4,6-dichloro-1H-pyrazolo[3,4-d]pyrimidine

  • MDL number MFCD10697159
  • Cas Number : 944902-17-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Referigrator
Physical Form Off white to Pale-yellow to Yellow-brown Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/41292.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/41292.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart