SY3H3D676DF6 Synthonix Corporation

4-Bromocyclohexanamine hydrochloride

  • MDL number MFCD14705051
  • Cas Number : 99337-81-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/41541.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/41541.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart