SY3H3D676DFF Synthonix Corporation

1-Bromo-2,4-dichloro-5-fluorobenzene

  • MDL number MFCD06659468
  • Cas Number : 1481-63-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/41634.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/41634.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart