SY3H3D677221 Synthonix Corporation

2-Bromo-1,4-dichlorobenzene

  • MDL number MFCD00018505
  • Cas Number : 1435-50-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Referigrator
Physical Form Crystal - Lump / Colour: White - Yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/41778.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/41778.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart