SY3H3D677232 Synthonix Corporation

6-Bromonicotinonitrile

  • MDL number MFCD00234141
  • Cas Number : 139585-70-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. refrigerator
Physical Form Crystal - Powder
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/41817.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/41817.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart